Anti-NOTCH1 Mouse Monoclonal Antibody [clone: 4C9]

Supplier: GeneTex
GTX84020
89366-784EA 1066.07 CAD
89366-784
Anti-NOTCH1 Mouse Monoclonal Antibody [clone: 4C9]
Antibodies
Mouse monoclonal antibody [4C9] to Notch1

Recommended Starting Dilutions:Western blot: 1:1000; IHC 1;50Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: NOTCH1
Clonality: Monoclonal
Clone: 4C9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR