Anti-Dengue Virus NS2B Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX124246
89359-378EA 903.1 CAD
89359-378
Anti-Dengue Virus NS2B Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Dengue Virus NS2B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Dengue virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR