Anti-WHSC1L1 Mouse Monoclonal Antibody [clone: 2E9]

Supplier: GeneTex

GTX50055
89363-686EA 657.02 CAD
89363-686
Anti-WHSC1L1 Mouse Monoclonal Antibody [clone: 2E9]
Antibodies
Mouse monoclonal antibody [2E9 ] to NSD3

We recommend the following starting dilutions: Western Blot: Use at 1:1,000 ~ 3,000.<p>Optimal working concentrations should be determined experimentally by the end user.

Type: Primary
Antigen: WHSC1L1
Clonality: Monoclonal
Clone: 2E9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR