Anti-NSF Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX110275
89356-252EA 673.08 CAD
89356-252
Anti-NSF Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to NSF (N-ethylmaleimide-sensitive factor)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: NSF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR