Anti-NSF Mouse Monoclonal Antibody [clone: NSF-1]

Supplier: GeneTex
GTX16681
89360-228EA 599.75 CAD
89360-228
Anti-NSF Mouse Monoclonal Antibody [clone: NSF-1]
Antibodies
N ethylmaleimide sensitive factor (NSF) is an ~83 kDa protein (predicted MW) which was initially isolated based on its ability to restore intercisternal Golgi transport to N ethylmaleimide treated membranes. NSF was subsequently discovered to be involved in a variety of membrane fusion events. NSF is an ATPase with two ATP binding domains that are essential for membrane fusion and homo oligomerization. In order to interact with target membranes, NSF requires another set of soluble proteins called Soluble NSF attachment proteins (SNAPs). NSF has been shown to play a prominent role in synaptic vesicle fusion. Together with synaptotagmin and a SNAP, NSF modulates the interaction of SNAP25, VAMP, and syntaxin, in an ATP dependent manner, to form a complex with a sedimentation coefficient of 20 S. This complex may represent an intermediate involved in synaptic vesicle docking and fusion.

IF: Use at an assay dependent dilution. IHC: Use at an assay dependent dilution. IP: Use at an assay dependent dilution. WB: Use at a dilution of 1/2000. Detects a band of approximately 76 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: NSF
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR