Anti-OCT4 Mouse Monoclonal Antibody [clone: 9B7]

Supplier: GeneTex
GTX83850
89366-780EA 1067.02 CAD
89366-780
Anti-OCT4 Mouse Monoclonal Antibody [clone: 9B7]
Antibodies
Mouse monoclonal antibody [9B7] to OCT4

Recommended Starting Dilutions:Western blot: 1:5000 ICC/IF (1:100)Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: OCT4
Clonality: Monoclonal
Clone: 9B7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR