Anti-RPA70 Mouse Monoclonal Antibody

GTX70261
89364-828EA 443.06 CAD
89364-828
Anti-RPA70 Mouse Monoclonal Antibody
Replication Protein A (RPA) is a single-stranded DNA binding protein. Human RPA is a heterotrimeric protein containing subunits of 70, 32 and 14 kDa. This protein complex is highly conserved in eukaryotes and is essential in DNA replication, homologous recombination and nucleotide excision repair. RPA has been shown to be a target for DNA-dependent protein kinase (DNA-PK) ataxia telangiectasia-mutated gene (ATM) and a cyclin dependent protein kinase. The middle subunit, RPA-32, is phosphorylated during the S-phase of the cell cycle, in response to DNA damage and during apoptotsis of Jurkat T- lymphocytes.

Type: Primary
Antigen: RPA70
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR