Anti-CDNK1A Mouse Monoclonal Antibody

GTX20118
89360-780EA 536.98 CAD
89360-780
Anti-CDNK1A Mouse Monoclonal Antibody
Useful as a detection system for rapidly dividing cells.

Type: Primary
Antigen: CDNK1A
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR