Anti-P21/RAS Rabbit Monoclonal Antibody

Supplier: Genetex
GTX61187
89363-866EA 585.93 CAD
89363-866
Anti-P21/RAS Rabbit Monoclonal Antibody
Ras, a signal transducer, was first characterized as the transforming genes of Harvey & Kristen sarcoma virus . The Ras family (H-Ras, N-Ras, and K-Ras) regulates cell growth, differentiation and apoptosis . Switching from an active or resting state, Ras can either bind GTP or GDP respectively. In the triphosphate conformation, Ras will interact with GTPase activating protein (GAP) to increase its activity . Mutations in any of the three isoforms can convert these proteins into active oncogenes. Additionally, Ras mutations are found in 30 % of all human cancer .

Type: Primary
Antigen: P21/RAS
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR