Anti-P53 Mouse Monoclonal Antibody

GTX75246
89365-722EA 494.82 CAD
89365-722
Anti-P53 Mouse Monoclonal Antibody
Type: Primary
Antigen: P53
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: 0
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR