Anti-P53 Rabbit Monoclonal Antibody

GTX61000
89363-756EA 556.26 CAD
89363-756
Anti-P53 Rabbit Monoclonal Antibody
p53 acts as both a tumor-suppressor and transcription factor that, upon activation by DNA damage and other cellular stress signals, leads to the transcription of genes triggering cell-cycle arrest, apoptosis, and DNA repair . The gene for the nuclear phosphoprotein p53 is the most commonly mutated gene yet identified in human cancers . p53 is frequently mutated or inactivated in about 60% of cancers .

Type: Primary
Antigen: P53
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR