Anti-p63 Rabbit Polyclonal Antibody

GTX102426
89349-588EA 507.32 CAD
89349-588
Anti-p63 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to p63 (tumor protein p63)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: p63
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR