Anti-TP73 Rabbit Monoclonal Antibody

GTX61267
89363-902EA 556.26 CAD
89363-902
Anti-TP73 Rabbit Monoclonal Antibody
p73 is a transcription factor that shares a significant amino acid identity with p53 and p63 . p73 and p63 can both activate p53-responsive genes and induce apoptosis upon over- expression Unlike p53, p73 is not induced by DNA damage and is not a target for inactivation by viral oncoproteins . Similar to p63, p73 has an additional coding region in the C-terminus (not found in p53) that can undergo complex alternative splicing giving rise to 3 splice forms (α, β, and γ) . Activation of p73 requires the presence of functional c-Abl. It has also be found that mutant p53 can bind to and inactivate p73 .

Type: Primary
Antigen: TP73
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR