Anti-PAK-Interacting (PIP-1) Rabbit Polyclonal Antibody

GTX70441
89364-988EA 425.76 CAD
89364-988
Anti-PAK-Interacting (PIP-1) Rabbit Polyclonal Antibody
The family of p21-activated protein kinases (PAKs) is composed of serine-threonine kinases whose activity is regulated by the GTPases, Rac and Cdc42. PAKs have been implicated in the regulation of mitogen-activated protein cascades, cellular morphological and cytoskeletal changes, neurite outgrowth, and cell apoptosis. A novel human PAK-interacting protein, hPIP1, interacts with PAK1 and inhibits the Cdc42/Rac-stimulated kinase activity suggesting that hPIP1 is a negative regulator of PAK and PAK signaling pathways.

Type: Primary
Antigen: PAK-Interacting (PIP-1)
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR