Anti-PAN CADHERIN Mouse Monoclonal Antibody

GTX11342
89357-540EA 443.06 CAD
89357-540
Anti-PAN CADHERIN Mouse Monoclonal Antibody
Cadherins are calcium dependent cell adhesion molecules, which play important role in the growth and development of cells via the mechanisms of control of tissue architecture and the maintenance of tissue integrity. They are expressed in tissue specific manner and and are required for assembly of cells into solid tissue. Individual cadherin molecules are known to co-operate with each other to form a linear cell adhesion zipper. In adhesion junctions cadherins are bound to beta and gamma catenins which in turnbind to alpha catenin, an actin binding protein. Cadherins play important part in tumor invasion and metastasis.

Type: Primary
Antigen: PAN CADHERIN
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR