Anti-PAP Mouse Monoclonal Antibody [clone: SPM312]

GTX17793
89351-380EA 535.05 CAD
89351-380
Anti-PAP Mouse Monoclonal Antibody [clone: SPM312]
Prostatic acid phosphatase is one of the two antigenic markers of prostatic carcinoma, the other being prostate specific antigen. It belongs to the kallikrein family of serine proteases and is suggested to act as a hydrolase to split phospharyl choline in semen and as a transferase. This antibody does not inhibit the enzymatic activity of PSAP. It reacts with non-neoplastic adult and fetal prostatic glands, primary and metastatic prostatic carcinomas. It shows no cross reaction with other phosphatases.

Type: Primary
Antigen: PAP
Clonality: Monoclonal
Clone: SPM312
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR