Anti-CDH3 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX113648
89357-666EA 881.1 CAD
89357-666
Anti-CDH3 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to P-cadherin (cadherin-3)

Suggested starting dilutions are as follows: WB: 1:1000-1:10000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CDH3
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR