Anti-PCNP Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX112059
89356-960EA 673.08 CAD
89356-960
Anti-PCNP Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to PCNP (PEST proteolytic signal containing nuclear protein)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: PCNP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR