Anti-Pclo Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX30910
89363-014EA 854.18 CAD
89363-014
Anti-Pclo Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to Piccolo

For ICC: Use at a dilution of 1:200-1:500. For IHC-P: Use at an assay dependent dilution. For WB: Use at a dilution of 1:1000-1:5000 (ECL Detection). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Pclo
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR