Anti-SCUBE2 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX85173
89353-916EA 920.76 CAD
89353-916
Anti-SCUBE2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to SCUBE2

Type: Primary
Antigen: SCUBE2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR