Anti-SHC1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX24890
89361-572EA 920.44 CAD
89361-572
Anti-SHC1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to SHC ( phospho Y239 + Y240 )

Recommended Starting Dilutions:
For WB: Use at a concentration of 0.5-2 µg/ml.
For IHC-P: Use at a dilution of 1:10-1:50.
Optimal working dilutions/ concentrations should be determined experimentally by the end user.

Type: Primary
Antigen: SHC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR