Anti-PDE6A Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX30751
89362-984EA 895.56 CAD
89362-984
Anti-PDE6A Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PDE6A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR