Anti-PDGFRA Rabbit Monoclonal Antibody

GTX62496
89364-472EA 556.26 CAD
89364-472
Anti-PDGFRA Rabbit Monoclonal Antibody
PDGF Receptor alpha is a cell surface tyrosine kinase receptor for members of the platelet-derived growth factor family. These growth factors are mitogens for cells of mesenchymal origin. The identity of the growth factor bound to a receptor monomer determines whether the functional receptor is a homodimer or a heterodimer, composed of both platelet-derived growth factor receptor alpha and beta polypeptides. Studies in knockout mice, where homozygosity is lethal, indicate that the alpha form of the platelet-derived growth factor receptor is particularly important for kidney development. Mice heterozygous for the receptor exhibit defective kidney phenotypes. Moreover, the SH2 domain of Shf can bind to PDGF Receptor alpha at tyrosine 720 .

Type: Primary
Antigen: PDGFRA
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR