Anti-PDGFRB Rabbit Monoclonal Antibody

Supplier: Genetex
GTX61782
89364-184EA 585.93 CAD
89364-184
Anti-PDGFRB Rabbit Monoclonal Antibody
Human platelet-derived growth factor (PDGF) receptor, is a receptor tyrosine kinase whose activity induces various signalling cascades leading to cell growth and proliferation. Phosphorylation of Tyrosine 857 (pY857) in the catalytic loop of the PDGF Receptor beta has been shown to increase kinase activity. Two major isoforms exist, PDGF receptor-α and PDGF receptor-β. PDGF receptor-β can only bind two specific PDGF isoforms, PDGF-β and PDGF-Δ . Upon ligand binding, the receptor undergoes autophosphorylation, allowing binding and activation of cytoplasmic SH2 domain-containing signal transduction molecules such as PI kinase , Grb2, and Src, among others.

Type: Primary
Antigen: PDGFRB
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR