Anti-PDGFRA Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX25460
89361-642EA 599.81 CAD
89361-642
Anti-PDGFRA Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to PDGF Receptor alpha ( phospho Y754 )

WB: Use at a concentration of 0.35 - 1 ?g/ml. Detects a band of approximately 170-185 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: PDGFRA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR