Anti-PDK2 Rabbit Monoclonal Antibody

GTX61866
89364-222EA 556.26 CAD
89364-222
Anti-PDK2 Rabbit Monoclonal Antibody
A member of Pyruvate dehydrogenase kinase (PDK1, 2, 3, and 4), PDK2 is a serine kinase which is involved in the regulation of mammalian glucose metabolism. During aerobic energy metabolism, mitochondrial pyruvate dehydrogenase complex (PDC) catalyzes decarboxylation of pyruvate to form acetyl-CoA . PDK2 inhibits PDC thru phosphorylation of the E1 alpha subunit of PDC and inactivates the complex . Highly expressed in heart and skeletal muscles, PDK2 is also responsible for the phosphorylation of Akt at Ser473, which leads to phosphorylation by PDK1 on Thr308 on Akt and full Akt activation .

Type: Primary
Antigen: PDK2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR