Anti-PEPSIN Goat Polyclonal Antibody

GTX40732
89363-286EA 452.03 CAD
89363-286
Anti-PEPSIN Goat Polyclonal Antibody
Digestive enzyme with particularly broad specificity.

Type: Primary
Antigen: PEPSIN
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Pig (Porcine, Swine)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR