Anti-Pyy Mouse Monoclonal Antibody [clone: RPY-B12]

Supplier: GeneTex
GTX59710
89363-734EA 1000.91 CAD
89363-734
Anti-Pyy Mouse Monoclonal Antibody [clone: RPY-B12]
Antibodies
Mouse monoclonal antibody [RPY-B12] to Peptide YY (PYY)

The antibody can be used for indirect ELISA (0.5 µg/ml) and Immunofluorescence.

Type: Primary
Antigen: Pyy
Clonality: Monoclonal
Clone: RPY-B12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR