Anti-PCNT Goat Polyclonal Antibody

Supplier: Genetex
GTX19044
89360-542EA 919.07 CAD
89360-542
Anti-PCNT Goat Polyclonal Antibody
Antibodies
Pericentrin is probably a conserved integral component of the filamentous matrix of the centrosome involved in the initial establishment of organized microtubule arrays.

Approx 75kDa band observed in HeLa lysates (calculated MW of 75kDa according to NP_079120). Recommended concentration: 1-3µg/ml. Please note that, in contrast to other anti-Pericentrin antibodies available, this ab is NOT expected to recognize Kendrin (also known as Pericentrin 2). This may explain why it recognizes a 75KDa band (the expected size according to NP_079120) and not a 220kDa band (as reported for other anti-pericentrin antibodies).

Type: Primary
Antigen: PCNT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR