Anti-PGCD Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX14786
89360-064EA 917.81 CAD
89360-064
Anti-PGCD Rabbit Polyclonal Antibody
Antibodies
In response to G-protein couples receptor stimulation, the cGMP can be produced from GTP by either cytoplasmic, soluble guanylate cyclase (sGC) are heterodimers ('N & 'O polypeptide chains), that are stimulated by nitric oxide and carbon monoxide or by particulate membrane-bound guanylyl cyclases which are activated by a complex mechanism by natriuretic peptides. Particulate GC (PGCs) have 7 different isoforms, PGC-A through PGC-G and are expressed in most tissues in isoform specific manner. There is significant structural homology among various PGCs, there is a large N-terminal extracellular domain (ECD), a single TMD and a large intracellular domain with protein kinase activity (KLD), a C-terminal catalytic domain (CD) and in between is a dimmerization domain (DD). The particulate GC-D, also known as olfactory gunylate cyclas, is specifically expressed in a subpopulation of olfactory sensory neurons.

WB: Use at a dilution of 1/500. Detects a band of approximately 135 kDa (predicted molecular weight: 120 kDa). A ~135 kDa band was seen using peptide controls and also when using rat olfactory bulb homogenates. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: PGCD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR