Anti-PLCG1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX24828
89361-534EA 737.64 CAD
89361-534
Anti-PLCG1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Phospholipase C1 gamma (phospho Tyr783)

WB: Use at a dilution of 1/1000. Predicted molecular weight: 135 kDa. Can be blocked with GTX25249 Phospholipase C gamma 1 phospho Y783 and non-phospho peptides (pair) or GTX25354 NIH3T3 whole cell lysate, + / - PDGF stimulation (pair) Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: PLCG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR