Anti-T Mouse Monoclonal Antibody [clone: 107.1]

Supplier: GeneTex
GTX72089
89352-950EA 565.77 CAD
89352-950
Anti-T Mouse Monoclonal Antibody [clone: 107.1]
Antibodies
Type: Primary
Antigen: T
Clonality: Monoclonal
Clone: 107.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR