Anti-PHOX2A Rabbit Polyclonal Antibody

GTX12046
89359-128EA 465.17 CAD
89359-128
Anti-PHOX2A Rabbit Polyclonal Antibody
Phox2a (paired-like homeobox 2a, aristaless homeobox homolog, Arix) is a transcription factor expressed in noradrenergic cell types of the sympathetic nervous system.

Type: Primary
Antigen: PHOX2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR