Anti-PI 3 Kinase P55 gamma Mouse Monoclonal Antibody

GTX20253
89360-816EA 513.69 CAD
89360-816
Anti-PI 3 Kinase P55 gamma Mouse Monoclonal Antibody
3-phosphorylates the cellular phosphoinositide PtdIns-4,5-biphosphate (PtdIns(4,5)P2) to produce PtdIns-3, 4,5-triiphosphate (PtdIns(3,4,5)P3). Links G-protein coupled receptor activation to the secondary messenger PtdIns(3,4,5)P3 production.

Type: Primary
Antigen: PI 3 Kinase P55 gamma
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Cow (Bovine)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR