Anti-IgA Goat Polyclonal Antibody

Supplier: GeneTex
GTX80646
89353-860EA 346.44 CAD
89353-860
Anti-IgA Goat Polyclonal Antibody
Antibodies
Goats were immunized with Pig IgA from normal pig serum. After multiple immunizations in Freund’s adjuvant serum was collected. The resulting antiserum was solid phase absorbed, immunoaffinity purified off an antigen containing immunoabsorbant then conjugated with horseradish (HRP).

Type: Primary
Antigen: IgA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR