Anti-PKC alpha Rabbit Monoclonal Antibody

GTX61959
89364-248EA 556.26 CAD
89364-248
Anti-PKC alpha Rabbit Monoclonal Antibody
Activation of PKC plays a crucial role in tranducing signals for a variety of cellular responses, including tumor proliferation . PKC isoforms have been classified into three groups: classical, novel, and atypical. Members of these three PKC groups contain an autoinhibitory domain that binds to the substrate binding site in the catalytic domain, preventing its activation in the absence of cofactors or activators . The activity of PKC is under the control of three distinct phosphorylation events, including that of the Thr641 autophosphorylation site .

Type: Primary
Antigen: PKC alpha (Phospho THR497)
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR