Anti-Plasminogen Goat Polyclonal Antibody

GTX11523
89358-286EA 562.83 CAD
89358-286
Anti-Plasminogen Goat Polyclonal Antibody
Plasminogen is a plasma protein synthesised mostly in the liver. Plasma concentration is usually in the range of 70 - 200mg/L. Plasminogen is activated by tissue-plasminogen activator, urokinase and streptokinase, to form plasmin. Activation results from the cleavage and release of the preactivation peptide. Angiostatin, an internal fragment of plasminogen, is a potent inhibitor of angiogenesis, which selectively inhibits endothelial cell proliferation. Angiostatin potently inhibits tumor growth and can maintain metastatic and primary tumors in a dormant state. It is defined by a balance of proliferation and apoptosis of the tumor cells and is composed of plasminogen first four cringle structures. This molecule is generated by proteolytic cleavage of plasminogen.

Type: Primary
Antigen: Plasminogen
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR