Anti-PLG Mouse Monoclonal Antibody [clone: 9D8]

Supplier: GeneTex
GTX10175
89350-662EA 762.63 CAD
89350-662
Anti-PLG Mouse Monoclonal Antibody [clone: 9D8]
Antibodies
Mouse monoclonal [9D8] to Plasminogen

AP: Use at an assay dependent dilution. ELISA: Use at an assay dependent dilution. IHC-P: Use at an assay dependent dilution. Antigen retrieval is not essential but may optimise staining. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: PLG
Clonality: Monoclonal
Clone: 9D8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR