Anti-POT1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX30601
89352-710EA 904.69 CAD
89352-710
Anti-POT1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to POT1

Uses and Dilutions: By Western blot a band at ~71.4 kDa is seen. No other applications have been tested. Recommended Starting Dilutions:*Western Blot: 1:500-1:2,000 (ECL) ** Immunoprecipitation: Not testedImmunohisto/cytochemistry: Not tested *Optimal working dilutions should be determined by the investigator. **Nuclear extracts may be necessary for optimal results. ***Nitrocellulose membranes may be necessary for optimal results, although this has not been completely determined.

Type: Primary
Antigen: POT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR