Anti-PPP2CA Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX106334
89354-628EA 640.65 CAD
89354-628
Anti-PPP2CA Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to PP2A alpha (protein phosphatase 2, catalytic subunit, alpha isozyme)

Suggested starting dilutions are as follows: WB: 1:1000-1:10000, IHC-P: 1:100-1:1000, ICC/IF: 1:100-1:1000, IP: 1:100-1:500. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: PPP2CA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR