Anti-PENK Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX80743
89366-676EA 934.08 CAD
89366-676
Anti-PENK Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PENK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR