Anti-PPIF Chicken Polyclonal Antibody

GTX82309
89368-066EA 642.96 CAD
89368-066
Anti-PPIF Chicken Polyclonal Antibody
Chicken polyclonal to Peptidyl-prolyl cis-trans isomerase. mitochondrial

ELISA (Direct-Only tested on protein): Affi-pure IgY as primary antibody (1 ug/ml) and Goat anti-IgY HRP as 2nd antibody. WB (only tested on fusion protein): Use at a dilution of 1:2000. Colorimetric method for signal development.

Type: Primary
Antigen: PPIF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR