Anti-INS Mouse Monoclonal Antibody [clone: 3A1]

Supplier: GeneTex

GTX28301
89352-422EA 533.93 CAD
89352-422
Anti-INS Mouse Monoclonal Antibody [clone: 3A1]
Antibodies
Mouse monoclonal [3A1] to Proinsulin

This antibody can be used in a two-site enzyme immunoassay with anti-insulin GTX28302 (7F8) for labeling. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: INS
Clonality: Monoclonal
Clone: 3A1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR