Anti-PRL Rabbit Polyclonal Antibody

GTX28554
89352-494EA 539.63 CAD
89352-494
Anti-PRL Rabbit Polyclonal Antibody
Rabbit polyclonal to Prolactin

For IHC-Fr: Use at a dilution of 1:20-1:30. For IHC-P: Use at a dilution of 1:20-1:30. It does not require heat mediated antigen retrieval. Prolonged fixation in buffered formalin can destroy the epitope. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: PRL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR