Anti-PSA Rabbit Monoclonal Antibody

GTX61579
89364-060EA 556.26 CAD
89364-060
Anti-PSA Rabbit Monoclonal Antibody
Prostate-specific antigen (PSA, Kallikrein-3, KLK3) is a serine protease that is usually secreted from the prostate gland and presented in seminal plasma. PSA function in the liquefaction of seminal coagulum, through hydrolysis of the high molecular mass seminal vesicle protein. Its substrates includes semenogelin I & II, fibronectin, insulin-like growth factor binding protein 3 (IGBP3), and laminin . Highly expressed by normal and malignant prostate cancer cells, PSA has been implicated in the growth, invasion, and metastasis of prostate cancer cells . Therefore, PSA is widely used as a diagnostic marker for prostate cancer. Recently, PSA has been suggested as an endothelial cell-specific inhibitor of angiogenesis .

Type: Primary
Antigen: PSA
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR