Anti-PSA Mouse Monoclonal Antibody [clone: 2H9]

Supplier: GeneTex
GTX10184
89350-670EA 1147.57 CAD
89350-670
Anti-PSA Mouse Monoclonal Antibody [clone: 2H9]
Antibodies
Mouse monoclonal [2H9] to PSA

ELISA: Use at an assay dependent dilution. Specificity in ELISA - equimolar total PSA, epitope 3. Detection of PSA antigen: use as an antibody pair with GTX10185. For the detection of free PSA: use as an antibody pair with GTX10187. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: PSA
Clonality: Monoclonal
Clone: 2H9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR