Anti-PSD2 Rabbit Polyclonal Antibody

GTX105942
89354-454EA 258.54 CAD
89354-454
Anti-PSD2 Rabbit Polyclonal Antibody
Type: Primary
Antigen: PSD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: 0
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR