Anti-DLG4 Rabbit Monoclonal Antibody

GTX61948
89364-242EA 585.93 CAD
89364-242
Anti-DLG4 Rabbit Monoclonal Antibody
Postsynaptic Density protein 95 (PSD95) belongs to the membrane-associated guanylate kinase (MAGUK) family. As a scaffolding protein, PSD95 controls of receptor trafficking during synaptic plasticity by anchoring synaptic proteins. Specifically, PSD95 is necessary in synaptic plasticity associated with NMDA receptor signaling . After hetermultimerization with DLG2, PSD95 associates with cytoplasmic tail of NMDA receptor subunit and potassium channel clusters. PSD95 has been linked to autism, drug addiction, and learning

Type: Primary
Antigen: DLG4
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR