Anti-PSTAIR Mouse Monoclonal Antibody

GTX10345
89350-004EA 491.37 CAD
89350-004
Anti-PSTAIR Mouse Monoclonal Antibody
The cell cycle is controlled in part by cyclin-dependent kinases (CDKs) which are themselves controlled by modifications such as phosphorylation of CDKs and formation of complex(es) with other proteins, including the cyclins. CDKs are key regulators of cell cycle progression. CDKs are closely related in size (35-40 kD) and sequence (>40% identity) and associate with and are activated by a cyclin which acts as a regulatory subunit.

Type: Primary
Antigen: PSTAIR
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR