Anti-PTGER3 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX13381
89359-658EA 717.21 CAD
89359-658
Anti-PTGER3 Rabbit Polyclonal Antibody
Antibodies
Prostaglandin E Receptor EP3 is a member of the G protein coupled receptor family. This protein is one of four receptors identified for prostaglandin E2 (PGE2). This receptor may have many biological functions, which involve digestion, nervous system, kidney reabsorption, and uterine contraction activities. Studies of the mouse counterpart suggest that this receptor may also mediate adrenocorticotropic hormone response as well as fever generation in response to exogenous and endogenous stimuli. Prostaglandin E2 Receptor EP3 expression has been documented throughout the periphery, especially kidney. ESTs have been isolated primarily from kidney libraries.

IHC-P: Use at a dilution of 1/100 - 1/200. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: PTGER3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR